Toluene-[D₈] (99.5% D) for NMR spectroscopy

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Phenylmethane-D8, Methylbenzene-D8, Toluene-[²H₈]

87159.0010EA 132 GBP
87159.0010
Toluene-[D₈] (99.5% D) for NMR spectroscopy
Toluene-[D8]
Formula: C₆D₅CD₃
MW: 100.08 g/mol
Boiling Pt: 110
Melting Pt: –85 °C
Density: 0.94 g/cm³ (20 °C)
Flash Pt: 4 °C (closed cup)
Storage Temperature: Refrigerator
MDL Number: MFCD00044638
CAS Number: 2037-26-5
EINECS: 218-009-5
UN: 1294
ADR: 3 II

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.9 %
Isotopic enrichment (FT NMR 400 MHz)(D) Min. 99.5 %
Water (HDO+D2O) Max. 0.0200 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR