Acetone-[D₆] Isotope_(99.8% D) NMR

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Propanone-D6, DMK-D6, Hexadeuteroacetone, Dimethyl ketone-D6, Acetone-[²H₆], Acetone-d6

87152.0010EA 54.6 GBP
87152.0010 87152.0100 87152.0006 87152.0011
Acetone-[D₆] Isotope_(99.8% D) NMR
Acetone-[D6]
Formula: D₃CC(O)CD₃
MW: 64.03 g/mol
Boiling Pt: 55.5 °C (1013 hPa)
Melting Pt: –95.4 °C
Density: 0.79 g/cm³ (20 °C)
Flash Pt: –19 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00044635
CAS Number: 666-52-4
EINECS: 211-563-9
UN: 1090
ADR: 3 II

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.9 %
Isotopic enrichment (FT NMR 400 MHz)(D) Min. 99.80 %
Water (HDO+D2O) Max. 0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR