(±)-2-Methyltetrahydrofuran ≥99.0% stabilised, GPR RECTAPUR® for synthesis

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


Stabilisation: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 150-400 ppm

87132.290EA 103.8 GBP
87132.290 87132.320 87132.460 87132.550
(±)-2-Methyltetrahydrofuran ≥99.0% stabilised, GPR RECTAPUR® for synthesis
(±)-2-Methyltetrahydrofuran
Formula: C₅H₁₀O
Boiling Pt: 79 °C (1013 hPa)
Melting Pt: –136 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: –12 °C (closed cup)
MDL Number: MFCD00005367
CAS Number: 96-47-9
EINECS: 202-507-4
UN: 2536
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colourless liquid
Assay Min. 99.0 %
IR Spectrum Passes test
Water Max. 0.03 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR