VWR®, Potassium standard solution, 1,000 mg/L K in dilute nitric acid AVS TITRINORM standard for AAS
Supplier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
86686.260EA 71.7 GBP
86686.260 86686.180
VWR®, Potassium standard solution, 1,000 mg/L K in dilute nitric acid AVS TITRINORM standard for AAS
Standards Cationic Standards Potassium Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%