Phenolphthalein, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. indicator

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

86158.150EA 25.5 GBP
86158.150 86158.180 86158.260
Phenolphthalein, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. indicator
Phenolphthalein
Formula: C₂₀H₁₄O₄
MW: 318.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005913
CAS Number: 77-09-8
Merck Index: 14,07243
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

pH range (Colourless to Violet-Red) 8 - 10
Absorption Maximum λ (pH 9.8) 551 - 554
Absorptivity (A1%/1 cm; λ max; pH 9.8 on Dried Sample) 500 - 600
Clarity of alcohol solution passes test
Loss on Drying (110 ºC) ≤1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR