Sodium dihydrogen phosphate, anhydrous ≥99%, HiPerSolv CHROMANORM® for HPLC

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

85916.150EA 85.2 GBP
85916.150 85916.230
Sodium dihydrogen phosphate, anhydrous ≥99%, HiPerSolv CHROMANORM® for HPLC
Sodium dihydrogen phosphate
Formula: NaH₂PO₄
Melting Pt: ∼650 °C
Density: 0.9996 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003527
CAS Number: 7558-80-7
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 12,08806

Order Now

Specification Test Results

Assay ≥ 99 %
Appearance White crystalline powder
Filter test Conforms to standard
RP gradient test Conforms to standard
Solubility in water (0.1 g/ml; 25°C) Colourless, complete
pH (20°C; 10 %) 4,0 - 4,5
Absorbance (210 nm) (0.5 mol/l) ≤ 0,1
Absorbance (260 nm) (0.5 mol/l) ≤ 0,06
Absorbance (280 nm) (0.5 mol/l) ≤ 0,04
Absorbance (320 nm) (0.5 mol/l) ≤ 0,02
Absorbance (500 nm) (0.5 mol/l) ≤ 0,02


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR