Chloroform, Wasser_0,005 Stabilised

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

Stabilisation: 0.005 % max. 2-Methyl-2-buten

85815.230EA 45.9 GBP
85815.230 85815.269
Chloroform, Wasser_0,005 Stabilised
Chloroform
Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 61.7 °C (1013 hPa)
Melting Pt: –63 °C
Density: 1.4832 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1 III
REACH: 01-2119486657-20
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC, on anhydrous basis) 99.9-100 %
Appearance Clear colorless liquid
Acidity (as HCl) 0-0.001 %
Residue after evaporation 0-0.0005 % w/w
Stabilizer (Amylene) 0.003-0.006 % w/w
Water (KF) 0-0.005 % w/w


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR