VWR®, Refractive index standard kits for calibration according to Ph. Eur. 2.2.6
Supplier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
85757.001EA 328 GBP
85757.001 85758.001
VWR®, Refractive index standard kits for calibration according to Ph. Eur. 2.2.6
Standards Refractive Index Standards
These standards are for the calibration of refractometers prior to use.