VWR® Buffer, Ammonium Chloride, pH 10, Reag. Ph.Eur.
Supplier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
85739.290EA 80 GBP
85739.290
VWR® Buffer, Ammonium Chloride, pH 10, Reag. Ph.Eur.
Buffers pH Calibration Buffers
The reagent is specially intended for use in testing Ph Eur and monographs. This solution was produced according 4.1.1 of Ph Eur specifications using Ph Eur reagent quality components and water meeting the Ph Eur monograph requirement for purified water.
  • Ph Eur N 4007300
Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.