Quaternary ammonium test strips, Dosatest, VWR®
Supplier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
85711.601EA 38.2 GBP
85711.601
Quaternary ammonium test strips, Dosatest, VWR®
Environmental Test Kits
Semi-quantitative test strips for the determination of QUAT.
  • Ideal for rapid and easy analysis of QUAT ammonium directly on-site
  • No maintenance or calibration is required
Colour change: Yellow  to  blue-green
Delivery information: Delivered as 100 test strips in a tube with instruction leaflet.