VWR® ARISTAR®, Zirconium standard solutions for ICP-MS
Supplier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
85649.180EA 79.2 GBP
85649.180 457483C
VWR® ARISTAR®, Zirconium standard solutions for ICP-MS
Standards Cationic Standards Zirconium Standards
  • Produced from high purity acids, water ASTM I 18 Megaohms
  • Solution assayed by titration
  • Final concentration verified against reference material from NIST
  • Total maximum uncertainty: 0,5% (typical)
  • Delivered with complete certificate of analysis
Certifications: Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities