VWR® ARISTAR®, Tellurium Standard Solutions for ICP-MS

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

85634.180EA 132 GBP
85634.180 457461A
VWR® ARISTAR®, Tellurium Standard Solutions for ICP-MS
Standards Cationic Standards Tellurium Standards

  • Produced from high purity acids, water ASTM I 18 Megaohms
  • Solution assayed by titration
  • Final concentration verified against reference material from NIST
  • Total maximum uncertainty: 0.5% (typical)
  • Delivered with complete certificate of analysis


Certifications: Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR