Span® 80, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ100

85548-1L 85548-250ML
85548-1LEA 108 GBP
85548-1L 85548-250ML
Span® 80, Supelco®
Span® 80
Formula: C₂₄H₄₄O₆
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00080948
CAS Number: 1338-43-8
EINECS: 215-665-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b4e0e15db043c16004bf42281caef11d69d6ee2&vid=36f9f073c049fe722daee943b718a4da79a0e276&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR