Acetonitrile, anhydrous (max. 0.001% H₂O) ≥99.95% for DNA synthesis

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


Filtered through a 0.2 µm filter, packaged under nitrogen.

85501.290EA 150 GBP
85501.290 85501.400 85501.320 85501.500 85501.911 85501.914 VWRC85501.321
Acetonitrile, anhydrous (max. 0.001% H₂O) ≥99.95% for DNA synthesis
Acetonitrile
Formula: H₃CCN
Boiling Pt: 81.6 °C (1013 hPa)
Melting Pt: –45.7 °C
Density: 0.782 g/cm³ (20 °C)
Flash Pt: 5 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99.95 %
IR Spectrum Passes test
Acidity Max. 0.001 meq/g
Alkalinity Max. 0.0002 meq/g
Colouration Max. 10 APHA
Evaporation residue Max. 1 ppm
Water Max. 10 ppm

SPECIFICATIONS

Environmentally Preferable Safer Human and Environmental Health Safer Human and Environmental Health Safer Human and Environmental Health Safer Human and Environmental Health Safer Human and Environmental Health
Environmental certification Chem21 Solvent Selection Guide recommended by default Chem21 Solvent Selection Guide recommended by default and after discussion EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion Chem21 Solvent Selection Guide recommended by default and after discussion EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR