Barium chloride 0.1 mol/l (0.2N) aqueous solution, AVS TITRINORM Volumetric

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

85496.230EA 27.8 GBP
85496.230
Barium chloride 0.1 mol/l (0.2N) aqueous solution, AVS TITRINORM Volumetric
Barium chloride
Formula: BaCl₂
MW: 208.24 g/mol
Boiling Pt: 1560
Melting Pt: 963
Storage Temperature: Ambient
MDL Number: MFCD00003445
CAS Number: 10361-37-2
Merck Index: 12,00998

Order Now

Specification Test Results

Titer (20°C; real value 0.2 % accuracy) 0,0998 -> 0,1002 mol/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR