Zinc sulphate 0.05 mol/l (0.05 N) aqueous solution USP Test Solution (TS)

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

85378.180EA 22.7 GBP
85378.180 85378.260
Zinc sulphate 0.05 mol/l (0.05 N) aqueous solution USP Test Solution (TS)
Zinc sulphate

Ready to use solutions.


  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available

Formula: O₄SZn
MW: 161.45 g/mol
Melting Pt: 100
Density: 1.45
Storage Temperature: Ambient
MDL Number: MFCD00011302
CAS Number: 7733-02-0
EINECS: 0
Merck Index: 12,10293

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR