Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution USP Test Solution (TS)

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

85374.180EA 29.9 GBP
85374.180 85374.260
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution USP Test Solution (TS)
Sodium thiosulphate

Ready to use test solutions (TS)


  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available

Formula: Na₂S₂O₃
Melting Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 0
Merck Index: 12,08844

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR