VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

85042.001EA 116 GBP
85042.001 85041.001 85043.001 85044.001 85045.001 85046.001 85041.002 85042.002 85043.002 85044.002 85045.002 85046.002
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Buffers pH Calibration Buffers

For calibration of pH meters.


  • Practical colour-coded 20 ml and 30 ml sachets - single-use per sachet - no risk of contamination
  • Ideal for on-site analyses
  • Traceable to SRM from NIST


Packaging: Two versions available: 30× 30 ml in a standard box and 30× 20 ml in a user-friendly dispenser box

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR