Ammonia solution 20%, NORMATOM®, Ultrapure for trace metal analysis

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

85031.270EA 436 GBP
85031.270 85031.230
Ammonia solution 20%, NORMATOM®, Ultrapure for trace metal analysis
Ammonia solution
Formula: NH₃
Density: 0.923 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8,III

Order Now

Specification Test Results

Assay 20-22%
Ag (Silver) Max. 10 ppt
Al (Aluminium) Max. 20 ppt
As (Arsenic) Max. 10 ppt
Au (Gold) Max. 10 ppt
Ba (Barium) Max. 10 ppt
Be (Beryllium) Max. 10 ppt
Bi (Bismuth) Max. 10 ppt
Ca (Calcium) Max. 10 ppt
Cd (Cadmium) Max. 10 ppt
Ce (Cerium) Max. 10 ppt
Co (Cobalt) Max. 10 ppt
Cr (Chromium) Max. 10 ppt
Cs (Cesium) Max. 10 ppt
Cu (Copper) Max. 10 ppt
Dy (Dysprosium) Max. 10 ppt
Er (Erbium) Max. 10 ppt
Eu (Europium) Max. 10 ppt
Fe (Iron) Max. 10 ppt
Ga (Gallium) Max. 10 ppt
Gd (Gadolinium) Max. 10 ppt
Ge (Germanium) Max. 10 ppt
Hf (Hafnium) Information only
Hg (Mercury) Max. 200 ppt
Ho (Holmium) Max. 10 ppt
In (Indium) Max. 10 ppt
K (Potassium) Max. 10 ppt
La (Lanthanum) Max. 10 ppt
Li (Lithium) Max. 10 ppt
Lu (Lutetium) Max. 10 ppt
Mg (Magnesium) Max. 10 ppt
Mn (Manganese) Max. 10 ppt
Mo (Molybdenum) Max. 10 ppt
Na (Sodium) Max. 20 ppt
Nb (Niobium) Max. 10 ppt
Nd (Neodymium) Max. 10 ppt
Ni (Nickel) Max. 10 ppt
Pb (Lead) Max. 10 ppt
Pd (Palladium) Information only
Pr (Praseodymium) Max. 10 ppt
Pt (Platinum) Information only
Rb (Rubidium) Max. 10 ppt
Re (Rhenium) Information only
Rh (Rhodium) Max. 10 ppt
Ru (Ruthenium) Information only
Sb (Antimony) Max. 10 ppt
Sc (Scandium) Max. 10 ppt
Se (Selenium) Information only
Sm (Samarium) Max. 10 ppt
Sn (Tin) Max. 10 ppt
Sr (Strontium) Max. 10 ppt
Tb (Terbium) Max. 10 ppt
Te (Tellurium) Max. 10 ppt
Th (Thorium) Max. 10 ppt
Ti (Titanium) Max. 10 ppt
Tl (Thallium) Max. 10 ppt
Tm (Thulium) Max. 10 ppt
U (Uranium) Max. 10 ppt
V (Vanadium) Max. 10 ppt
W (Tungsten) Max. 10 ppt
Y (Yttrium) Max. 10 ppt
Yb (Ytterbium) Max. 10 ppt
Zn (Zinc) Max. 10 ppt
Zr (Zirconium) Max. 10 ppt


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR