VWR®, Ammonium (in N) 1000 mg/L in water standard for Ion Chromatography
Supplier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
84953.180EA 66.5 GBP
84953.180 84953.260
VWR®, Ammonium (in N) 1000 mg/L in water standard for Ion Chromatography
Standards Cationic Standards Ammonium Standards
Certifications: Traceable to NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities. Supplied with Certificate of Analysis.