Ammonium acetate ≥99%, HiPerSolv CHROMANORM® for LC-MS

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

84885.180EA 106 GBP
84885.180 84885.260
Ammonium acetate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Ammonium acetate
Formula: H₃CCOONH₄
Melting Pt: 114 °C
Density: 1.17 g/cm³ (20 °C)
Flash Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD00013066
CAS Number: 631-61-8
EINECS: 211-162-9
Merck Index: 13,00495
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99 %
Ignition residue Max. 0.01 %
Water Max. 2 %
Cl (Chloride) Max. 5 ppm
SO4 (Sulphate) Max. 10 ppm
NO3 (Nitrate) Max. 10 ppm
Ca (Calcium) (0.1 %) Max. 0.2 ppm
K (Potassium) (0.1 %) Max. 0.2 ppm
Mg (Magnesium) (0.1 %) Max. 0.2 ppm
Na (Sodium) (0.1 %) Max. 0.5 ppm
Fe (Iron) (0.1%) Max. 0.2 ppm
Transmittance (210 nm) (0.1 %) Min. 10 %
Transmittance (220 nm) (0.1 %) Min. 50 %
Transmittance (230 nm) (0.1 %) Min. 80 %
Transmittance (235 nm) (0.1 %) Min. 95 %
Transmittance (245 nm) (0.1 %) Min. 99 %
Suitable for LC-MS Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR