VWR®, UV/Visible spectroscopy stray light standards
Supplier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
84784.180EA 171 GBP
84784.180 84789.180
VWR®, UV/Visible spectroscopy stray light standards
Standards Colour Reference and Spectroscopy Standards
Stray light standards packed in permanently sealed UV cuvettes or 100 ml amber bottle.