VWR® Buffer Solutions, Secondary Standards
Supplier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
84582.260EA 57.5 GBP
84582.260 84583.260 84580.260 84581.260 84584.260 84585.260 84586.260 84587.260 84593.260
VWR® Buffer Solutions, Secondary Standards
Buffers pH Calibration Buffers
  • Accurate to ±0,010 pH units (except buffer 12,454 with accuracy ±0,050)
  • Solution standardised at 25 °C
  • Fully traceable for optimum pH calibration (traceable to NIST, DIN 19266 and IUPAC)
  • Temperature dependence data is printed on the labels as are batch number and expiry date