Hydrochloric acid 4 M HCl water, Fluka™

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

84435-1L
84435-1LEA 37.1 GBP
84435-1L
Hydrochloric acid 4 M HCl water, Fluka™
Hydrochloric acid

Honeywell's high-quality titration products cover the complete titration workflow and include:


  • Solutions for volumetric titration (acids, bases, or salts)Buffers available as concentrates or ready-to-useReagents for complexometry:Aminopolycarboxylic acids (EDTA/NTA analogs)
  • Masking agents
  • Indicators for volumetric and complexometric titrations


UNSPSC Code: 12000000

Formula: HCl
MW: 36.46 g/mol
Density: 1.072 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II

Order Now

Specification Test Results

Appearance (color) complying
Appearance (form) complying
Assay 3.95 - 4.10 mol/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR