Acetone 99,9, PESTINORM® GC_capillary

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

Filter 0,2 um_Nitrogen_filled

83960.320EA 98.8 GBP
83960.320 83960.400
Acetone 99,9, PESTINORM® GC_capillary
Acetone
Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.2 °C (1013 hPa)
Melting Pt: –95.4 °C
Density: 0.792 g/cm³ (20 °C)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3 II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.9 %
Acidity Max. 0.0005 meq/g
Evaporation residue (100°C) Max. 0.0005 %
Organic residue (as Octanol) (GC/FID) Max. 10 ng/ml
Halogenated residue (as Lindane)(GC/ECD) Max. 5 ng/l
Water Max. 0.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR