Ammonium iron (III) citrate, TECHNICAL

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

83887.290EA 116 GBP
83887.290 83887.360
Ammonium iron (III) citrate, TECHNICAL
Ammonium iron (III) citrate
Formula: C₆H₅+4y)Fe(x)N(y)O₇
Storage Temperature: Ambient
MDL Number: MFCD00013099
CAS Number: 1185-57-5
EINECS: 214-686-6

Order Now

Specification Test Results

Assay (calculated as Fe) 14 - 16 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR