n-Hexane ≥95%, PESTINORM® for pesticide residue analysis

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

Filtered through a 0.2 µm filter, packaged under nitrogen.

83661.320EA 148 GBP
83661.320 83661.290 83661.400
n-Hexane ≥95%, PESTINORM® for pesticide residue analysis
n-Hexane
Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69
Melting Pt: –94,3 °C
Density: 0.659 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
REACH: 01-2119480412-44
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Limited Time Sale

Select items below are currently on sale

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 95,0%
Evaporation residue Max. 5 ppm
Water Max. 0,02 %
Pesticide analysis (Lindane/ECD) Max. 5 ng/l

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR