Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for HPLC

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Acetic acid ethyl ester

Filtered through a 0.2 µm filter, packaged under nitrogen.

83621.320EA 146 GBP
83621.320 83621.290 83621.9010 83621.9030
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Ethyl acetate
Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77.1 °C (1013 hPa)
Melting Pt: –83 °C
Density: 0.902 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3 II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.8 %
Acidity Max. 0.0005 meq/g
Alkalinity Max. 0.0002 meq/g
Evaporation residue Max. 5 ppm
Water Max. 0.1 %
Absorbance (260 nm) Max. 0.15
Absorbance (270 nm) Max. 0.05
Absorbance (300 nm) Max. 0.02
Transmittance (260 nm) Min. 70 %
Transmittance (270 nm) Min. 90 %
Transmittance (300 nm) Min. 96 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR