Yttrium(III) fluoride ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Yttrium trifluoride, Yttrium(III) fluoride

83106.14 83106.22
83106.14EA 57.2 GBP
83106.14 83106.22
Yttrium(III) fluoride ≥98%
Yttrium (III) fluoride

Purity: 98% CAS No.: 13709-49-4 Molecular Formula: F3Y Formula Weight: 145.90 Physical Form: -325 Mesh Powder Sensitivity: Hygroscopic Appearance: White Melting Point: 1387° UN No.: 3288 MDL No.: MFCD00049714

Formula: F₃Y
Boiling Pt: 2227 °C (1013 hPa)
Melting Pt: 1387 °C
Density: 4.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049714
CAS Number: 13709-49-4
EINECS: 237-257-5
UN: 3288
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR