Petroleum spirit, 100…140 °C, BAKER, Laboratory Reagent, J.T.Baker®

Supplier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

8243.5000EA 148 GBP
8243.5000 8243.9025 8243.1000 8243.9200
Petroleum spirit, 100…140 °C, BAKER, Laboratory Reagent, J.T.Baker®
Petroleum spirit
Boiling Pt: 100…140 °C (1013 hPa)
Melting Pt: –40 °C
Density: 0.728 g/cm³ (15 °C)
Flash Pt: 1 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3,II

Order Now

Specification Test Results

Boiling Range 100 - 140 °C
Residue after Evaporation max. 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR