Petroleum spirit, 40…60 °C, BAKER, Laboratory Reagent, J.T.Baker®

Supplier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

8240.1000EA 44.8 GBP
8240.1000 8240.5000 8240.9025 8240.9200
Petroleum spirit, 40…60 °C, BAKER, Laboratory Reagent, J.T.Baker®
Petroleum spirit
Boiling Pt: 40…60 °C (1013 hPa)
Density: 0.65 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3,II

Order Now

Specification Test Results

Appearance passes test
Appearance at 0°C passes test
Distillation Range 40 - 65 °C
Residue after Evaporation max. 0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR