VWR®, Cleanroom Spiral Notebooks
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
818-0279EA 208 GBP
818-0279 818-0278 818-0280
VWR®, Cleanroom Spiral Notebooks
Notebooks
These cleanroom spiral notebooks are made of synthetic latex paper.
  • White
  • 100 pages
  • Polypropylene cover
  • Spiral bound
  • 85 gsm
Suitable for cleanroom use.