n-Pentane, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Supplier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Pentane

8114.2500EA 238 GBP
8114.2500 8114.1000 8114.9025 8114.9200
n-Pentane, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
n-Pentane
Formula: H₃C(CH₂)₃CH₃
Boiling Pt: 35…36 °C (1013 hPa)
Melting Pt: –130 °C
Density: 0.63 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3,II
REACH: 01-2119459286-30
Merck Index: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (by GC) min. 99 %
Boiling Range 35 - 37 °C
Density (g/ml) at 20°C 0.624 - 0.626
Residue after Evaporation max. 0.001 %
Sulfur Compounds (as S) max. 0.005 %
Water (H2O) max. 0.01 %
Trace Impurities (in ppm)Aluminium (Al) max. 0.5
Trace Impurities (in ppm)Boron (B) max. 0.02
Trace Impurities (in ppm)Barium (Ba) max. 0.1
Trace Impurities (in ppm)Calcium (Ca) max. 0.5
Trace Impurities (in ppm)Cadmium (Cd) max. 0.05
Trace Impurities (in ppm)Cobalt (Co) max. 0.02
Trace Impurities (in ppm)Chromium (Cr) max. 0.02
Trace Impurities (in ppm)Copper (Cu) max. 0.02
Trace Impurities (in ppm)Iron (Fe) max. 0.1
Trace Impurities (in ppm)Magnesium (Mg) max. 0.1
Trace Impurities (in ppm)Manganese (Mn) max. 0.02
Trace Impurities (in ppm)Nickel (Ni) max. 0.02
Trace Impurities (in ppm)Lead (Pb) max. 0.1
Trace Impurities (in ppm)Tin (Sn) max. 0.1
Trace Impurities (in ppm)Zinc (Zn) max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR