Lab markers
Supplier: MICRONOVA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
PEN-40 PEN-40IR
811-0177EA 232 GBP
811-0177 811-0178
Lab markers
Pens Lab Markers
LabMarkers feature permanent ink that marks on cold or wet surfaces, plastic bags or disposable labware. Marks will not smear on reusable glass, metal or porcelain labware but can be scrubbed off.
  • Fine tip, black ink
  • Permanent on most surfaces
  • Fast drying
  • Available gamma irradiated
Materials of construction: High-density poly barrel. Permanent, quick drying, alcohol- resistant black ink.
Chemical compatibility: Holds up to wipedown with IPA, bleach, and phenols. Integrity of printed barrel will be compromised with thinners and strong solvents.