Cleanroom markers, Sharpie®
Supplier: MICRONOVA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
PEN-27IR PEN-26IR PEN-22IRDB PEN-23IRDB PEN-20IRDB
811-0176EA 185 GBP
811-0176 811-0175 811-0198 811-0199 811-0197
Cleanroom markers, Sharpie®
Pens Cleanroom Pens
The industry standard, original pen style, fine point, permanent marker.
  • Marks permanently on most surfaces
  • Fade and water resistant ink
  • Quick drying, non toxic alcohol-based ink formula
Sharpie® fine point markers write on cold or wet surfaces, plastic bags or disposable labware. Marks will not smear on reusable glass, metal, or porcelain labware but can be scrubbed off. Sterile items are gamma irradiated and suitable for Aseptic ISO 5 and other clean production areas.
Packaging: Double bagged, gamma irradiated.