Pyridine, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Azabenzene, Azine

8073.1000EA 209 GBP
8073.1000 8073.2500
Pyridine, BAKER ANALYZED® ACS, J.T.Baker®
Pyridine
Formula: C₅H₅N
Boiling Pt: 115 °C (1013 hPa)
Melting Pt: –44 °C
Density: 0.983 g/cm³ (20 °C)
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
EINECS: 203-809-9
UN: 1282
ADR: 3,II
Merck Index: 13,08060
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f9c5934940218027dae0d80a1f98b6d5179c0a8&vid=9f2ef46a26e488d366e82869b3acb96652b5dab1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Exceeds ACS Specifications
Assay (by GC) min. 99.0 %
Ammonia (NH3) max. 0.002 %
Chloride (Cl) max. 0.001 %
Reducing Substances passes test
Residue after Evaporation max. 0.002 %
Solubility in Water passes test
Suitability for OH groups determ. by phthalisation passes test
Sulfate (SO4) max. 0.001 %
Water (H2O) max. 0.1 %
Trace Impurities (in ppm)
Aluminium (Al) max. 0.5
Boron (B) max. 0.1
Barium (Ba) max. 0.1
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.05
Cobalt (Co) max. 0.02
Chromium (Cr) max. 0.02
Copper (Cu) max. 0.02
Iron (Fe) max. 0.1
Magnesium (Mg) max. 0.1
Manganese (Mn) max. 0.02
Nickel (Ni) max. 0.02
Lead (Pb) max. 0.1
Tin (Sn) max. 0.1
Zinc (Zn) max. 1.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR