Beakers, tall form, DURAN®
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
211171705 211172401 211172907 211173603 211174102 211174805 211175407
213-0253EA 9.6 GBP
213-0253 213-1190 801-3129 801-3136 801-3141 215-1012 801-3154
Beakers, tall form, DURAN®
Beakers
DURAN®, borosilicate glass 3.3, clear, without spout.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Graduated for approximate reading of contents and labelling area.
With printed Retrace Code (batch identification).
Certifications: ISO 3819