Ethanol ≥96% (v/v), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

8007.1000EA 34.3 GBP
8007.1000 8007.5000 8007.2500 8007.2500PE 8007.1000DP BAKR8007.5000_DP
Ethanol ≥96% (v/v), BAKER ANALYZED® ACS, J.T.Baker®
Ethanol
Formula: C₂H₅OH
Melting Pt: –114 °C
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II

Order Now

Specification Test Results

Exceeds ACS Specifications
Assay 95.1 - 96.9 % (v/v)
Acetone, isopropylalcohol passes test
Color (APHA) max. 10
Methanol (CH3OH) max. 0.1 %
Residue after Evaporation max. 0.001 %
Solubility in Water passes test
Substances Darkened by H2SO4 passes test
Substances Reducing KMnO4 passes test
Titrable Acid (meq/g) max. 0.0005
Titrable Base (meq/g) max. 0.0002
Trace Impurities (in ppm)
Aluminium (Al) max. 0.5
Boron (B) max. 0.02
Barium (Ba) max. 0.1
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.05
Cobalt (Co) max. 0.02
Chromium (Cr) max. 0.02
Copper (Cu) max. 0.02
Iron (Fe) max. 0.1
Magnesium (Mg) max. 0.1
Manganese (Mn) max. 0.02
Nickel (Ni) max. 0.02
Lead (Pb) max. 0.1
Tin (Sn) max. 0.1
Zinc (Zn) max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR