Copper(I) cyanide for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Copper cyanide, Cuprous cyanide

M-Clarity™ quality level = MQ200

8.41811.0100 8.41811.1000 8.41811.0500
8.41811.0100EA 35.7 GBP
8.41811.0100 8.41811.1000 8.41811.0500
Copper(I) cyanide for synthesis, Sigma-Aldrich®
Copper (I) cyanide
Formula: CCuN
Melting Pt: 474 °C
Density: 2.92 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010975
CAS Number: 544-92-3
EINECS: 208-883-6
UN: 1587
ADR: 6.1,II
Merck Index: 13,02691

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point 473 °C
Molar Mass 89.56 g/mol
Bulk Density 750 kg/m3
Density 2.92 g/cm3
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR