α,α,α-Trifluoro-p-cresol for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning


M-Clarity™ quality level = MQ200

8.41244.0001 8.41244.0005
8.41244.0001EA 46.9 GBP
8.41244.0001 8.41244.0005
α,α,α-Trifluoro-p-cresol for synthesis, Sigma-Aldrich®
α,α,α-Trifluoro-p-cresol
Formula: C₇H₅F₃O
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 45…47 °C
Density: 1.3 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002363
CAS Number: 402-45-9
EINECS: 206-945-7

Order Now

Specification Test Results

Melting Point 46 - 48 °C
Molar Mass 162.11 g/mol
Boiling Point 73 - 75 °C (11 hPa)
Flash Point 83 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR