Ethyl-4-(dimethylamino)benzoate for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 4-(Dimethylamino)benzoic acid ethyl ester, Ethyl 4-dimethylaminobenzoate

M-Clarity™ quality level = MQ200

8.41086.0100 8.41086.9999
8.41086.0100EA 86.5 GBP
8.41086.0100 8.41086.9999
Ethyl-4-(dimethylamino)benzoate for synthesis, Sigma-Aldrich®
Ethyl 4-(dimethylamino)benzoate
Formula: C₁₁H₁₅NO₂
MW: 193.25 g/mol
Boiling Pt: 112 °C (0.02 mmHg)
Melting Pt: 63…65 °C
Density: 1.0099 g/cm³ (99.7 °C)
MDL Number: MFCD00009115
CAS Number: 10287-53-3
EINECS: 233-634-3

Order Now

Specification Test Results

Melting Point 63 - 65 °C
Molar Mass 193.24 g/mol
Synonyms 4-Dimethylaminobenzoic acid ethyl ester
Hill Formula C11H15NO2

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR