Sodium-1-decanesulphonate for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Sodium decane-1-sulphonate, Decane-1-sulphonic acid sodium salt, 1-Decanesulphonic acid, sodium salt

M-Clarity™ quality level = MQ200

8.41036.0010 8.41036.0050 8.41036.0002
8.41036.0010EA 87.8 GBP
8.41036.0010 8.41036.0050 8.41036.0002
Sodium-1-decanesulphonate for synthesis, Sigma-Aldrich®
Sodium 1-decanesulphonate
Formula: C₁₀H₂₁NaO₃S
MW: 244.33 g/mol
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007526
CAS Number: 13419-61-9
EINECS: 236-525-9

Order Now

Specification Test Results

Melting Point 300 °C
Molar Mass 244.33 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR