4-Aminobenzoic acid for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Vitamin Bx, para-Aminobenzoic acid, PABA, Vitamin H1

M-Clarity™ quality level = MQ200

8.22312.0250 8.22312.1000 8.22312.9025
8.22312.0250EA 49.9 GBP
8.22312.0250 8.22312.1000 8.22312.9025
4-Aminobenzoic acid for synthesis, Sigma-Aldrich®
4-Aminobenzoic acid
Formula: C₇H₇NO₂
Boiling Pt: 340 °C (1013 hPa)
Melting Pt: 188 °C
Density: 1.45 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00007894
CAS Number: 150-13-0
EINECS: 205-753-0
Merck Index: 13,00422

Order Now

Specification Test Results

Solubility 4.7 g/l (20 °C)
Melting Point 186 - 189 °C
Molar Mass 137.14 g/mol
Bulk Density 200 kg/m3
Density 1.38 g/cm3
pH 3.5 (5 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR