Ethyl-(S)-(-)-lactate for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Ethyl (S)-(-)-2-hydroxypropionate, Ethyl L(-)-lactate, (S)-Ethyl 2-hydroxypropanoate, (S)-(-)-Ethyl lactate

M-Clarity™ quality level = MQ200

8.22100.0100 8.22100.1000 8.22100.9025
8.22100.0100EA 34 GBP
8.22100.0100 8.22100.1000 8.22100.9025
Ethyl-(S)-(-)-lactate for synthesis, Sigma-Aldrich®
Ethyl (S)-(-)-lactate
Formula: C₅H₁₀O₃
MW: 118.13 g/mol
Boiling Pt: 154
Melting Pt: –25 °C
Density: 1.042 g/cm³ (20 °C)
Flash Pt: 53
Storage Temperature: Ambient
MDL Number: MFCD00004518
CAS Number: 687-47-8
EINECS: 211-694-1
UN: 1192
ADR: 3 III
Merck Index: 12,03850

Order Now

Specification Test Results

Solubility (25 °C) soluble
Melting Point -25 °C
Molar Mass 118.13 g/mol
Boiling Point 154 °C (1013 hPa)
Vapor Pressure 1.6 hPa (20 °C)
Flash Point 53 °C
Refractive Index 1.411 (25 °C, 589 nm)
Explosion Limit 1.5 - 16.4 %(V)
Density 1.03 g/cm3 (20 °C)
pH 4 (50 g/l, H2O, 20 °C)
Ignition Point 400 °C
Viscosity kinematic 2.7 mm2/s (25 °C)
Hazard Class 3
Hazard Class Description Flammable Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR