2-Chlorobenzoyl chloride for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ200

8.21989.0250
8.21989.0250EA 45 GBP
8.21989.0250
2-Chlorobenzoyl chloride for synthesis, Sigma-Aldrich®
2-Chlorobenzoyl chloride
Formula: C₇H₄Cl₂O
Boiling Pt: 238 °C (1013 hPa)
Melting Pt: –4 °C
Density: 1.38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000660
CAS Number: 609-65-4
EINECS: 210-194-0
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Melting Point -4 °C
Molar Mass 175.01 g/mol
Boiling Point 225 °C (1013 hPa)
Vapor Pressure 0.1 hPa (20 °C)
Flash Point 124 °C
Refractive Index 1.5718 (20 °C, 589 nm)
Explosion Limit 1.5 - 9.4 %(V)
Density 1.38 g/cm3 (20 °C)
pH (H2O) acidic
Ignition Point 600 °C
Viscosity kinematic 1.89 mm2/s (20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR