Pyrazinamide for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Pyrazinecarboxamide, Pyrazine-2-carboxamide, 2-Pyrazinecarboxamide

M-Clarity™ quality level = MQ200

8.21050.0025
8.21050.0025EA 51.8 GBP
8.21050.0025
Pyrazinamide for synthesis, Sigma-Aldrich®
Pyrazinamide
Formula: C₅H₅N₃O
MW: 123.11 g/mol
Boiling Pt: 357
Melting Pt: 188…191 °C
MDL Number: MFCD00006132
CAS Number: 98-96-4
EINECS: 202-717-6
Merck Index: 12,08140

Order Now

Specification Test Results

Hill Formula C₅H₅N₃O
Molar Mass 123.12 g/mol
Melting Point 188 - 191 °C
pH value 7 (H₂O)
Solubility (20 °C) slightly soluble
Storage class 10 - 13 Other liquids and solids
WGK WGK 2 water endangering


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR