Polyvinyl alcohol, M.W. ∼ 30000 zS, hydrolysed_fully, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Poly(vinyl alcohol), PVOH, PVA, Mowiol®

MQ200

8.21039.0100 8.21039.1000
8.21039.0100EA 27.8 GBP
8.21039.0100 8.21039.1000
Polyvinyl alcohol, M.W. ∼ 30000 zS, hydrolysed_fully, Sigma-Aldrich®
Polyvinyl alcohol
Formula: [-CH₂CH(OH)-]n
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now

Specification Test Results

Solubility Soluble at 20 °C
Density 1.3 g/cm3 (20 °C)
pH 4.5 - 7 (40 g/l, H2O, 20 °C)
Ignition Point 450 °C
pH value 3,5 - 7,0
Storage temperature +15...+25 °C
Loss on drying ≤5,0 %
Degree of hydrolysis ≥98%
Dynamic viscosity 3,8 - 5,2 mPa.s
Ash ≤1,0 %
Ester value 12 - 25


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR