Phenothiazine for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning


M-Clarity™ quality level = MQ200

8.20975.0100 8.20975.1000 8.20975.9050
8.20975.0100EA 36.4 GBP
8.20975.0100 8.20975.1000 8.20975.9050
Phenothiazine for synthesis, Sigma-Aldrich®
Phenothiazine
Formula: C₁₂H₉NS
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 182…185 °C
Density: 1.34 g/cm³ (20 °C)
Flash Pt: 202 °C
Storage Temperature: Ambient
MDL Number: MFCD00005015
CAS Number: 92-84-2
EINECS: 202-196-5
UN: 3077
ADR: 9,III
Merck Index: 12,07404

Order Now

Specification Test Results

Solubility  - 0.001 g/l (20 °C)
Melting Point 182 - 185 °C
Molar Mass 199.26 g/mol
Bulk Density 650 kg/m3
Boiling Point 381 °C (1011 hPa)
Vapor Pressure  - 0.001 hPa (30 °C)
Density 1.34 g/cm3 (20 °C)
pH 6 (10 g/l, H2O, 20 °C) (aqueous suspension)
Ignition Point 470 °C
Hill Formula C12H9NS


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR