4-(4-Nitrobenzyl)pyridine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 4-(p-Nitrobenzyl)Pyridine

M-Clarity™ quality level = MQ200

8.20890.0025 8.20890.0100 8.20890.0005
8.20890.0025EA 138 GBP
8.20890.0025 8.20890.0100 8.20890.0005
4-(4-Nitrobenzyl)pyridine, Sigma-Aldrich®
4-(4-Nitrobenzyl)pyridine
Formula: C₁₂H₁₀N₂O₂
MW: 214.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006445
CAS Number: 1083-48-3
EINECS: 214-108-2

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 68 - 71 °C
Molar Mass 214.22 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR