2-Imidazolidinethione for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 2-Mercaptoimidazoline, Ethlenethiourea, Ethylenethiourea, 2-Imidazolidinethione

M-Clarity™ quality level = MQ200

8.20696.0250 8.20696.1000 8.20696.9025
8.20696.0250EA 48.7 GBP
8.20696.0250 8.20696.1000 8.20696.9025
2-Imidazolidinethione for synthesis, Sigma-Aldrich®
2-Imidazolidinethione
Formula: C₃H₆N₂S
Storage Temperature: Ambient
MDL Number: MFCD00005276
CAS Number: 96-45-7
EINECS: 202-506-9

Order Now

Specification Test Results

Solubility 8 g/l (20 °C)
Melting Point 196 - 199 °C
Molar Mass 102.16 g/mol
Bulk Density 750 - 900 kg/m3
Vapor Pressure 0.2 hPa (170 °C)
Flash Point 253 °C
Density 1.4 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR